EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Measuring level alignment at the metal-molecule interface by in situ electrochemical (13) C NMR.
PMID 25639536
Abstract
A new technique to measure energy-level alignment at a metal-molecule interface between the Fermi level of the metal and the frontier orbitals of the molecule is proposed and experimentally demonstrated. The method, which combines the electrochemistry of organo-ligand-stabilized Au nanoparticles with (13) C NMR spectroscopy (i.e. in situ electrochemical NMR), enables measuring both occupied and unoccupied states.