EMAIL THIS PAGE TO A FRIEND
Journal of the Egyptian Society of Parasitology
Assessment of diagnostic and therapeutic approaches of Helicobacter pylori-associated iron deficiency and anemia in children with dyspeptic symptoms.
PMID 25643511
Abstract
This study assessed the diagnostic approaches of Helicobacter pylori (IP)-associated iron deficiency (ID) and anemia (IDA) in children with dyspeptic symptoms and evaluated the effect of simultaneous anti-H. pylori (anti-HIP) therapy and oral iron in comparison with each of anti? HP therapy and oral iron therapy alone, on iron status as assessed by serum soluble transferrin receptor (sTfR) level. Two hundreds children with dyspeptic symptoms were subjected to clinical evaluation, stool examination, CBC, biochemical assays for serum iron parameters and measurements of serum IgG antibodies to HP and serum sTfR level by ELISA. Sixty children were found to have HP. associated ID or IDA and were randomly divided into 3 groups (20 children each). GA received 2-week anti-HP therapy plus 90-day oral iron, and GB received 2-week anti-HP therapy alone whereas group C received 90-day oral iron alone. Re-evaluation of the 3 groups was performed after 3 months of treatment initiation by repeat CBC and serum sTfR level. Children (45%) were HP-seropositive. The mean values of serum sTfR were significantly higher in HP-positive group and in HP-positive children with IDA than in HP-negative group and in HP-negative children with IDA although no significant differences were noted in hematologic variables and iron parameters between the corresponding groups and children. As regard treatment groups, there were significant improvements in the mean values of indices of IDA status (HIb, MCH, MCV, sTfR) and ID status (sTtRi) at 3 months of treatment initiation compared with their baseline values after. anti-HP triple therapy either with oral iron or without oral iron whereas the control children who were treated with oral iron alone showed insignificant changes despite oral iron administration. The improvements in these parameters were significantly greater in groups of children who received anti-HP therapy either combined with iron or alone, where compared with those who did not receive anti-HP therapy. Their magnitudes were significantly higher among children receiving anti-HP therapy combined with oral iron when compared with that receiving anti-HP therapy alone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3518
Carbonyl iron, ≥97% Fe basis
Fe
F4503 Ferritin from equine spleen, Type I, saline solution
F6754 Ferritin from human liver, Type IV, 10 μg/mL
F6879 Ferritin from human spleen, Type V, 10 μg/mL in 0.15 M NaCl, 10 mM Tris, pH 8.0, containing 0.1% sodium azide
44890
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Fe
12310
Iron, ≥99%, reduced, powder (fine)
Fe
267945
Iron, chips, 99.98% trace metals basis
Fe
338141
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Fe
356808
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Fe
413054
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Fe
513423
Iron, activated, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 99.5% trace metals basis
Fe
IRMM524A
Iron, IRMM® certified Reference Material, 0.1 mm foil
Fe
IRMM524B
Iron, IRMM® certified Reference Material, 0.5 mm wire
Fe
255637
Iron, powder, ≥99.99% trace metals basis
Fe
209309
Iron, powder, −325 mesh, 97%
Fe
267953
Iron, powder, <10 μm, ≥99.9% trace metals basis
Fe
266213
Iron, rod, diam. 6.3 mm, 99.98% trace metals basis
Fe
266256
Iron, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Fe
266248
Iron, wire, diam. 0.5 mm, ≥99.99%
Fe
356824
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Fe
356832
Iron, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Fe
GF58945078
Iron, wire reel, 250m, diameter 0.25mm, hard, 99.5%
Fe
GF21695722
Iron, wire reel, 2m, diameter 0.13mm, hard, 99.99+%
Fe
GF81282135
Iron, wire reel, 2m, diameter 0.25mm, annealed, 99.99+%
Fe
GF57565996
Iron, wire reel, 2m, diameter 0.25mm, hard, 99.99+%
Fe
GF48022152
Iron, wire reel, 2m, diameter 0.5mm, hard, 99.99+%
Fe
GF48020947
Iron, wire reel, 500m, diameter 0.125mm, hard, 99.5%
Fe
GF58809965
Iron, wire reel, 50m, diameter 0.065mm, hard, 99.5%
Fe
GF34071566
Iron, wire reel, 50m, diameter 0.25mm, hard, 99.5%
Fe
GF96915134
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Fe