EMAIL THIS PAGE TO A FRIEND
Genes & development
Bifurcating action of Smoothened in Hedgehog signaling is mediated by Dlg5.
PMID 25644602
Abstract
Binding of the Hedgehog (Hh) protein signal to its receptor, Patched, induces accumulation of the seven-pass transmembrane protein Smoothened (Smo) within the primary cilium and of the zinc finger transcription factor Gli2 at the ciliary tip, resulting ultimately in Gli-mediated changes in nuclear gene expression. However, the mechanism by which pathway activation is communicated from Smo to Gli2 is not known. In an effort to elucidate this mechanism, we identified Dlg5 (Discs large, homolog 5) in a biochemical screen for proteins that preferentially interact with activated Smo. We found that disruption of Smo-Dlg5 interactions or depletion of endogenous Dlg5 leads to diminished Hh pathway response without a significant impact on Smo ciliary accumulation. We also found that Dlg5 is localized at the basal body, where it associates with another pathway component, Kif7. We show that Dlg5 is required for Hh-induced enrichment of Kif7 and Gli2 at the tip of the cilium but is dispensable for Gpr161 exit from the cilium and the consequent suppression of Gli3 processing into its repressor form. Our findings suggest a bifurcation of Smo activity in Hh response, with a Dlg5-independent arm for suppression of Gli repressor formation and a second arm involving Smo interaction with Dlg5 for Gli activation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU182641 MISSION® esiRNA, esiRNA targeting mouse Dlg5