EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Very low density lipoprotein receptor regulates dendritic spine formation in a RasGRF1/CaMKII dependent manner.
PMID 25644714
Abstract
Very Low Density Lipoprotein Receptor (VLDLR) is an apolipoprotein E receptor involved in synaptic plasticity, learning, and memory. However, it is unknown how VLDLR can regulate synaptic and cognitive function. In the present study, we found that VLDLR is present at the synapse both pre- and post-synaptically. Overexpression of VLDLR significantly increases, while knockdown of VLDLR decreases, dendritic spine number in primary hippocampal cultures. Additionally, knockdown of VLDLR significantly decreases synaptophysin puncta number while differentially regulating cell surface and total levels of glutamate receptor subunits. To identify the mechanism by which VLDLR induces these synaptic effects, we investigated whether VLDLR affects dendritic spine formation through the Ras signaling pathway, which is involved in spinogenesis and neurodegeneration. Interestingly, we found that VLDLR interacts with RasGRF1, a Ras effector, and knockdown of RasGRF1 blocks the effect of VLDLR on spinogenesis. Moreover, we found that VLDLR did not rescue the deficits induced by the absence of Ras signaling proteins CaMKIIα or CaMKIIβ. Taken together, our results suggest that VLDLR requires RasGRF1/CaMKII to alter dendritic spine formation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV42010
Anti-GCG antibody produced in rabbit, IgG fraction of antiserum
SAB2104861
Anti-GCG antibody produced in rabbit, affinity isolated antibody
SAB1410515
Anti-GCG antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501137
Anti-Glucagon antibody produced in rabbit, affinity isolated antibody
SAB4300405
Anti-GRIN1 (Ab-896) antibody produced in rabbit, affinity isolated antibody
SAB4300538
Anti-GRIN1 (Ab-897) antibody produced in rabbit, affinity isolated antibody
SAB4300425
Anti-GRIN2B (Ab-1474) antibody produced in rabbit, affinity isolated antibody
SAB2104208
Anti-GRIN2B antibody produced in rabbit, affinity isolated antibody
GW21015 Anti-HRAS antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405964
Anti-HRAS antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4501301
Anti-NMDAR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501302
Anti-NMDAR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501303
Anti-NMDAR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501305
Anti-NMDAR2B, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501306
Anti-NMDAR2B, C-Terminal antibody produced in rabbit, affinity isolated antibody
EHU091521 MISSION® esiRNA, esiRNA human VLDLR (esiRNA1)
EMU039901 MISSION® esiRNA, esiRNA targeting mouse Vldlr (esiRNA1)
WH0002641M1
Monoclonal Anti-GCG antibody produced in mouse, clone 2D3-2B11, purified immunoglobulin, buffered aqueous solution
SAB1403855
Monoclonal Anti-GCG antibody produced in mouse, clone 1E2-E6, purified immunoglobulin, buffered aqueous solution
SAB1402214
Monoclonal Anti-GRIN2B antibody produced in mouse, clone 2G5, purified immunoglobulin, buffered aqueous solution
SAB4100537
Monoclonal Anti-HRAS antibody produced in mouse, clone HRAS-A485, culture supernatant