EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacy and pharmacology
Investigation of the interactions of enteric and hydrophilic polymers to enhance dissolution of griseofulvin following hot melt extrusion processing.
PMID 25645386
Abstract
This study focuses on the application of hot melt extrusion (HME) to produce solid dispersions containing griseofulvin (GF) and investigates the in-vitro dissolution performance of HME powders and resulting tablet compositions containing HME-processed dispersions. Binary, ternary and quaternary dispersions containing GF, enteric polymer (Eudragit L100-55 or AQOAT-LF) and/or vinyl pyrrolidone-based polymer (Plasdone K-12 povidone or S-630 copovidone) were processed by HME. Two plasticizers, triethyl citrate (TEC) and acetyl tributyl citrate (ATBC), were incorporated to aid in melt processing and to modify release of GF in neutral media following a pH-change in dissolution. Products were characterized for GF recovery, degrees of compositional amorphous character, intermolecular interactions and non-sink dissolution performance. Binary dispersions exhibited lower maximum observed concentration values and magnitudes of supersaturated GF in neutral media dissolution in comparison with the ternary dispersions. The quaternary HME products, 1 : 2 : 1 : 0.6 GF : L100-55 : S-630 : ATBC and GF : AQOAT-LF : K-12 : ATBC, were determined as the most optimal concentration-enhancing compositions due to increased hydrogen bonding of enteric functional groups with carbonyl/acetate groups of vinyl pyrrolidone-based polymers, reduced compositional crystallinity and presence of incorporated hydrophobic plasticizer. HME products containing combinations of concentration-enhancing polymers can supersaturate and sustain GF dissolution to greater magnitudes in neutral media following the pH-transition and be compressed into immediate-release tablets exhibiting similar dissolution profiles.
Related Materials