EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
Dense fibrillar collagen is a potent inducer of invadopodia via a specific signaling network.
PMID 25646088
Abstract
Cell interactions with the extracellular matrix (ECM) can regulate multiple cellular activities and the matrix itself in dynamic, bidirectional processes. One such process is local proteolytic modification of the ECM. Invadopodia of tumor cells are actin-rich proteolytic protrusions that locally degrade matrix molecules and mediate invasion. We report that a novel high-density fibrillar collagen (HDFC) matrix is a potent inducer of invadopodia, both in carcinoma cell lines and in primary human fibroblasts. In carcinoma cells, HDFC matrix induced formation of invadopodia via a specific integrin signaling pathway that did not require growth factors or even altered gene and protein expression. In contrast, phosphoproteomics identified major changes in a complex phosphosignaling network with kindlin2 serine phosphorylation as a key regulatory element. This kindlin2-dependent signal transduction network was required for efficient induction of invadopodia on dense fibrillar collagen and for local degradation of collagen. This novel phosphosignaling mechanism regulates cell surface invadopodia via kindlin2 for local proteolytic remodeling of the ECM.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45983
Acetonitrile, analytical standard
C2H3N
PHR1551
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
00712
Acetonitrile solution, ~20% in H2O, for protein sequence analysis
C2H3N
SAB4500976
Anti-ARHGEF12 antibody produced in rabbit, affinity isolated antibody
HPA042689 Anti-ARHGEF18 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA013903 Anti-TIAM2 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
A6390
β-D-Allose, rare aldohexose sugar
C6H12O6
14339
Bicinchoninic acid disodium salt hydrate, ≥99.0% (HPLC)
C20H10N2Na2O4 · xH2O
1012772
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
C2H6O
PHR1070
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
C2H6O
E4009
Eosin Y, Dye content ~99 %
C20H8Br4O5
230251
Eosin Y
C20H8Br4O5
E6003
Eosin Y disodium salt, Dye content ≥85 %
C20H6Br4Na2O5
E4382
Eosin Y disodium salt, certified by the Biological Stain Commission
C20H6Br4Na2O5
45240
Eosin Y disodium salt, for microscopy (Fl., Hist.), adsorption and fluorescent indicator
C20H6Br4Na2O5
53019
Eosin Y disodium salt, analytical standard
C20H6Br4Na2O5
318906
Eosin Y solution, 5 wt. % in H2O
C20H6Br4Na2O5
49897
Formate standard for IC, 1.000 g/L in H2O, analytical standard
CH2O2
06473
Formic acid solution, BioUltra, 1.0 M in H2O
CH2O2
I1149
Iodoacetamide, BioUltra
C2H4INO
I6125
Iodoacetamide, ≥99% (NMR), crystalline
C2H4INO
I5161
Iodoacetamide, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production
C2H4INO
76523
L-Glutamine, certified reference material, TraceCERT®
C5H10N2O3
PHR1125
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
C5H10N2O3
C2456
Monoclonal Anti-Collagen, Type I antibody produced in mouse, clone COL-1, ascites fluid
PHR1320
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
1601340
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
C2H3N
1706698
Urea, United States Pharmacopeia (USP) Reference Standard
CH4N2O
U0600000
Urea, European Pharmacopoeia (EP) Reference Standard
CH4N2O
56180
Urea, analytical standard
CH4N2O