EMAIL THIS PAGE TO A FRIEND
Diabetes research and clinical practice
Changes in the TNF-alpha/IL-10 ratio in hyperglycemia-associated pregnancies.
PMID 25648390
Abstract
TNF-α is a diabetogenic cytokine associated with adverse outcomes during pregnancy that can be counterbalanced by IL-10. We have investigated IL-10 and TNF-α balance at maternal and placental levels in hyperglycemia-associated pregnancies. One hundred and ninety-two pregnant women participated, which included normoglycemic women (ND) and women with mild gestational hyperglycemia (MGH), gestational diabetes mellitus (GDM) and type 2 diabetes mellitus (DM2). Maternal plasma and placental tissue IL-10 and TNF-α levels were measured by ELISA and placental TNF-α was also immunolocalized. Maternal plasma TNF-α levels were highest in GDM (p=0.0190), whereas TNF-α levels were highest in placental tissues in DM2 (p=0.0095). Immunohistochemistry also showed strong reactivity with anti-TNF-α antibody in the villous structures in the DM2 group. Conversely, IL-10 levels were lowest in maternal plasma of the DM2 group (p=0.0228). The TNF-α/IL-10 ratio in maternal plasma progressively increased with the severity of hyperglycemia (p<0.0001), being highest in placenta of the DM2 group (p=0.0150). In both, plasma and placenta, TNF-α/IL-10 ratio were correlated with mean maternal glycemia and HbA1c levels. Alterations of placenta and serum TNF-α/IL-10 balance with predominance of TNF-α were correlated with the severity of hyperglycemia during gestation. This association may offer insight into the pathogenesis of gestational hyperglycemia and associated pregnancy outcomes.