EMAIL THIS PAGE TO A FRIEND
Marine drugs
Design, synthesis and biological evaluation of novel bromophenol derivatives incorporating indolin-2-one moiety as potential anticancer agents.
PMID 25648512
Abstract
A series of bromophenol derivatives containing indolin-2-one moiety were designed and evaluated that for their anticancer activities against A549, Bel7402, HepG2, HeLa and HCT116 cancer cell lines using MTT assay in vitro. Among them, seven compounds (4g-4i, 5h, 6d, 7a, 7b) showed potent activity against the tested five human cancer cell lines. Wound-healing assay demonstrated that compound 4g can be used as a potent compound for inactivating invasion and metastasis by inhibiting the migration of cancer cells. The structure-activity relationships (SARs) of bromophenol derivatives had been discussed, which were useful for exploring and developing bromophenol derivatives as novel anticancer drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
87920
Tetramethylsilane, analytical standard, for NMR spectroscopy, ACS reagent
C4H12Si