EMAIL THIS PAGE TO A FRIEND
American journal of obstetrics and gynecology
A description of the methods of the Nulliparous Pregnancy Outcomes Study: monitoring mothers-to-be (nuMoM2b).
PMID 25648779
Abstract
The primary aim of the "Nulliparous Pregnancy Outcomes Study: monitoring mothers-to-be" is to determine maternal characteristics, which include genetic, physiologic response to pregnancy, and environmental factors that predict adverse pregnancy outcomes. Nulliparous women in the first trimester of pregnancy were recruited into an observational cohort study. Participants were seen at 3 study visits during pregnancy and again at delivery. We collected data from in-clinic interviews, take-home surveys, clinical measurements, ultrasound studies, and chart abstractions. Maternal biospecimens (serum, plasma, urine, cervicovaginal fluid) at antepartum study visits and delivery specimens (placenta, umbilical cord, cord blood) were collected, processed, and stored. The primary outcome of the study was defined as pregnancy ending at <37+0 weeks' gestation. Key study hypotheses involve adverse pregnancy outcomes of spontaneous preterm birth, preeclampsia, and fetal growth restriction. We recruited 10,037 women to the study. Basic characteristics of the cohort at screening are reported. The "Nulliparous Pregnancy Outcomes Study: monitoring mothers-to-be" cohort study methods and procedures can help investigators when they plan future projects.