EMAIL THIS PAGE TO A FRIEND
Journal of the American Association for Laboratory Animal Science : JAALAS
Dose regimens, variability, and complications associated with using repeat-bolus dosing to extend a surgical plane of anesthesia in laboratory mice.
PMID 25650976
Abstract
Extending a surgical plane of anesthesia in mice by using injectable anesthetics typically is accomplished by repeat-bolus dosing. We compared the safety and efficacy of redosing protocols administered either during an anesthetic surgical plane (maintaining a continuous surgical plane, CSP), or immediately after leaving this plane (interrupted surgical plane, ISP) in C57BL/6J mice. Anesthesia was induced with ketamine, xylazine, and acepromazine (80, 8, and 1 mg/kg IP, respectively), and redosing protocols included 25% (0.25K), 50% (0.5K), or 100% (1.0K) of the initial ketamine dose or 25% (0.25KX) or 50% (0.5KX) of the initial ketamine-xylazine dose. In the ISP group, the surgical plane was extended by 13.8 ± 2.1 min (mean ± SEM) after redosing for the 0.25K redose with 50% returning to a surgical plane, 42.7 ± 4.5 min for the 0.5K redose with 88% returning to a surgical plane, and 44.3 ± 15.4 min for the 1.0K redose, 52.8 ± 7.2 min for the 0.25KX redose, and 45.9 ± 2.9 min for the 0.5KX redose, with 100% of mice returning to a surgical plane of anesthesia in these 3 groups. Mortality rates for ISP groups were 0%, 12%, 33%, 12%, and 18%, respectively. Mice in CSP groups had 50% mortality, independent of the repeat-dosing protocol. We recommend redosing mice with either 50% of the initial ketamine dose or 25% of the initial ketamine-xylazine dose immediately upon return of the pedal withdrawal reflex to extend the surgical plane of anesthesia in mice, optimize the extension of the surgical plane, and minimize mortality.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7111
Acepromazine maleate, ≥98% (HPLC)
C19H22N2OS · C4H4O4
1001502
Acepromazine maleate, United States Pharmacopeia (USP) Reference Standard
C19H22N2OS · C4H4O4