EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Rational ligand design for the arylation of hindered primary amines guided by reaction progress kinetic analysis.
PMID 25651374
Abstract
We report the Pd-catalyzed arylation of very hindered α,α,α-trisubstituted primary amines. Kinetics-based mechanistic analysis and rational design have led to the development of two biarylphosphine ligands that allow the transformation to proceed with excellent efficiency. The process was effective in coupling a wide range of functionalized aryl and heteroaryl halides under mild conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
900275
(t-Bu)PhCPhos, 95%
C26H33N2P
900534
(t-Bu)PhCPhos Pd G3, ≥95%
C39H46N3O3PPdS