EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Biphasic Synthesis of Large-Pore and Well-Dispersed Benzene Bridged Mesoporous Organosilica Nanoparticles for Intracellular Protein Delivery.
PMID 25652950
Abstract
Large pore (4.6-7.6 nm) and well-dispersed benzene bridged mesoporous organosilica nanoparticles with uniform particle size of ≈50 nm are prepared via a biphasic approach. They can be directly used as nanocarriers without surface modification for the intracellular delivery of therapeutic proteins.