EMAIL THIS PAGE TO A FRIEND
Neurology
Antiepileptic drug use by pregnant women enrolled in Florida Medicaid.
PMID 25653296
Abstract
The study aims were to investigate secular trends in antiepileptic drug (AED) use in women during pregnancy, and to compare the use of first- and second-generation AEDs. Study participants consisted of female Florida Medicaid beneficiaries, older than 15 years, and pregnant within the time period 1999 to 2009. Fifteen AEDs were categorized into first and second generation of AEDs. Continuous use of AEDs was defined as at least 2 consecutive AED prescriptions totaling more than a 30-day supply. Polytherapy was defined as 2 or more AEDs continuously used for at least 30 overlapping days. Annual prevalence was estimated and compared. We included 2,099 pregnant women who were enrolled in Florida Medicaid from 1999 to 2009 and exposed to AEDs during pregnancy. Although there were fluctuations, overall AED use in the study cohort did not increase from 2000 to 2009 (β ± standard error [SE]: -0.07 ± 0.06, p = 0.31). The use of first-generation AEDs decreased (β ± SE: -6.21 ± 0.47, p < 0.0001), whereas the use of second-generation AEDs increased (β ± SE: 6.27 ± 0.52, p < 0.0001) from 2000 to 2009. AED use in polytherapy did not change through the study period. Valproate use reduced from 23% to 8% in the study population (β ± SE: -1.61 ± 0.36, p = 0.0019), but this decrease was only for women receiving an AED for epilepsy and was not present for other indications. The second-generation AEDs are replacing first-generation AEDs in both monotherapy and polytherapy. Valproate use has declined for epilepsy but not other indications. Additional changes in AED use are expected in future years.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
460036
5-Norbornene-2-endo,3-exo-dicarboxylic acid, 97%
C9H10O4