EMAIL THIS PAGE TO A FRIEND
Oncotarget
By downregulating TIAM1 expression, microRNA-329 suppresses gastric cancer invasion and growth.
PMID 25654811
Abstract
Gastric cancer (GC) is one of the most common malignant tumors worldwide. Emerging evidence has shown that abnormal microRNAs (miRNAs) expression is involved in tumorigenesis. MiR-329 was previously reported to act as a tumor suppressor or oncogene in some types of cancer. However, its function in gastric cancer (GC) is unclear. Here, we found that miR-329 was down-regulated in GC compared with adjacent controls. Enforced expression of miR-329 inhibited proliferation, migration and invasion of gastric cancer cells in vitro. We identified T lymphoma invasion and metastasis 1 (TIAM1) gene as potential target of miR-329. MiR-329 levels inversely correlated with TIAM1 expression in GC. Importantly, TIAM1 rescued the miR-329-mediated inhibition of cell invasion and proliferation. Finally, reintroduction of miR-329 significantly inhibited tumor formation of GC in the xenograft mice. Our findings suggest that miR-329 is a tumor suppressor and potential therapeutic target of GC.