EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Biomimetic microfingerprints for anti-counterfeiting strategies.
PMID 25656227
Abstract
An unclonable, fingerprint-mimicking anti-counterfeiting strategy is presented that encrypts polymeric particles with randomly generated silica film wrinkles. The generated wrinkle codes are as highly unique as human fingerprints and are technically irreproducible. Superior to previous physical unclonable functions, codes are tunable on demand and generable on various geometries. Reliable authentication of real-world products that have these microfingerprints is demonstrated using optical decoding methods.