EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Quantitative characterization of hydration state and destructuring effect of monosaccharides and disaccharides on water hydrogen bond network.
PMID 25658066
Abstract
Terahertz time-domain attenuated total reflection measurements of monosaccharide (glucose and fructose) and disaccharide (sucrose and trehalose) solutions from 0.146 M to 1.462 M were performed to evaluate (1) the hydration state and (2) the destructuring effect of saccharide solutes on the hydrogen bond (HB) network. Firstly, the extent of hydration water was determined by the decreased amount of bulk water with picosecond relaxation time that was replaced by that with much longer orientational relaxation time. As a result, we found glucose and trehalose exhibits stronger hydration capacity than fructose and sucrose, respectively, despite of the same number of the hydroxyl groups. For each saccharide, the hydration number tended to decrease with solute concentration. Secondly, the destructuring effect of these saccharide solutes on the HB network of the surrounding bulk water was discussed from the perspective of the fraction of non-hydrogen-bonded (NHB) water isolated from the HB network. We found the fraction of NHB water molecules that are not engaged in the HB network monotonously increased with saccharide concentration, indicating saccharide solutes promote the disruption of the water HB network. However, no noticeable differences were confirmed in the fraction of NHB water between glucose and fructose or between sucrose and trehalose. In contrast to hydration number, the number of NHB water produced by a single saccharide solute was less dependent on solute concentration, and three monosaccharide/disaccharide solutes were found to produce one/two NHB water molecules.