EMAIL THIS PAGE TO A FRIEND
Enzyme and microbial technology
Stability and activity of Dictyoglomus thermophilum GH11 xylanase and its disulphide mutant at high pressure and temperature.
PMID 25659634
Abstract
The functional properties of extremophilic Dictyoglomus thermophilum xylanase (XYNB) and the N-terminal disulphide-bridge mutant (XYNB-DS) were studied at high pressure and temperature. The enzymes were quite stable even at the pressure of 500MPa at 80°C. The half-life of inactivation in these conditions was over 30h. The inactivation at 80°C in atmospheric pressure was only 3-times slower. The increase of pressure up to 500MPa at 80°C decreased only slightly the enzyme's stability, whereas in 500MPa the increase of temperature from 22 to 80°C decreased significantly more the enzyme's stability. While the high temperature (80-100°C) decreased the enzyme reaction with short xylooligosaccharides (xylotetraose and xylotriose), the high pressure (100-300MPa) had an opposite effect. The temperature of 100°C strongly increased the Km but did not affect the kcat to the same extent, thus indicating that the interaction of the substrate with the active site suffers before the catalytic reaction begins to decrease as the temperature rises. Circular dichroism spectroscopy showed the high structural stability of XYNB and XYNB-DS at 93°C.