EMAIL THIS PAGE TO A FRIEND
Experimental eye research
Acetylated 8-oxoguanine DNA glycosylase 1 and its relationship with p300 and SIRT1 in lens epithelium cells from age-related cataract.
PMID 25660075
Abstract
The human 8-oxoguanine-DNA glycosylase 1 (OGG1) is the major DNA glycosylase responsible for repair of 7,8-dihydro-8-oxoguanine (8-oxoG) and ring-opened fapyguanine, critical mutagenic DNA lesions that are induced by reactive oxygen species. OGG1 acetylation has been demonstrated playing an important role in response to DNA damage. Here, we investigated the relationship between acetylated OGG1 (Ac-OGG1) and ARC, and clarified the effect of p300 and SIRT1 on the 8-oxoG excision ability of OGG1 in ARC development. Our results showed that anterior lens capsules from ARC group had higher proportion of 8-oxoG positive LECs than those from control group. OGG1 mRNA and protein levels significantly increased in ARC group compared with control group, while the protein levels of Ac-OGG1 were lower in ARC group. We investigated the factors involved in OGG1 acetylation and found that p300 and SIRT1 are the major acetyltransferases for OGG1 acetylation. We also identified acetylation of K338/K341 lysine residues in OGG1 has an important role on the repair activity of OGG1 to oxidative damage after H2O2 exposure in human lens epithelial cells (HLE-B3). Taken together, these data demonstrate that OGG1 acetylation regulates its function in response to DNA damage and could be one of the mechanisms of ARC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
660124
4,4′-Bis(N-carbazolyl)-1,1′-biphenyl, 97%
C36H24N2
699195
4,4′-Bis(N-carbazolyl)-1,1′-biphenyl, sublimed grade, 99.9% trace metals basis
C36H24N2
HPA003128
Anti-EP300 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004112
Anti-EP300 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300412
Anti-HDAC2 (Ab-394) antibody produced in rabbit, affinity isolated antibody
HPA011727
Anti-HDAC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV32618
Anti-HDAC2 antibody produced in rabbit, affinity isolated antibody
SAB2104633
Anti-HDAC2 antibody produced in rabbit, affinity isolated antibody
SAB4501384
Anti-HDAC2, C-Terminal antibody produced in rabbit, affinity isolated antibody
H3159
Anti-Histone Deacetylase 2 (HDAC2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
H2663
Anti-Histone Deacetylase 2 (HDAC2) antibody, Mouse monoclonal, clone HDAC2-62, purified from hybridoma cell culture
SAB4503485
Anti-p300, C-Terminal antibody produced in rabbit, affinity isolated antibody
P7493
Anti-PCAF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV32466
Anti-PCAF antibody produced in rabbit, IgG fraction of antiserum
SAB2101734
Anti-PCAF antibody produced in rabbit, affinity isolated antibody
WH0002033M2
Monoclonal Anti-EP300 antibody produced in mouse, clone 1D2, purified immunoglobulin, buffered aqueous solution
SAB1400094
Monoclonal Anti-EP300 antibody produced in mouse, clone 1B1, purified immunoglobulin, buffered aqueous solution
SAB1403776
Monoclonal Anti-EP300 antibody produced in mouse, clone 1D2, ascites fluid
WH0008850M2
Monoclonal Anti-PCAF antibody produced in mouse, clone 1H2, purified immunoglobulin, buffered aqueous solution
SAB1404637
Monoclonal Anti-PCAF antibody produced in mouse, clone 5E11, purified immunoglobulin, buffered aqueous solution
SAB1409953
Monoclonal Anti-PCAF antibody produced in mouse, clone 5E5, purified immunoglobulin, buffered aqueous solution
SAB1409954 Monoclonal Anti-PCAF antibody produced in mouse, clone 4C10, purified immunoglobulin, buffered aqueous solution
SAB1409955 Monoclonal Anti-PCAF antibody produced in mouse, clone 4D11, purified immunoglobulin, buffered aqueous solution
PLA0096
Rabbit anti-p300 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.