EMAIL THIS PAGE TO A FRIEND
Clinical endocrinology
Higher serum testosterone and dihydrotestosterone, but not oestradiol, are independently associated with favourable indices of lung function in community-dwelling men.
PMID 25660119
Abstract
Lower circulating androgens and poorer lung function are associated with increased cardiovascular risk and mortality in men. The association between androgens and lung function is unclear. We tested the hypothesis that circulating testosterone (T) and its metabolites dihydrotestosterone (DHT) and oestradiol (E2) are differentially associated with lung function in men. Early-morning serum T, DHT and E2 were assayed using mass spectrometry in 1768 community-dwelling men from Busselton, Western Australia. Forced expiratory volume in 1xa0s (FEV1) and forced vital capacity (FVC) were measured using spirometry. Linear regression models adjusting for age, height, smoking, exercise, body mass index, respiratory conditions and cardiovascular risk factors were used. Mean age was 50.1xa0±xa016·8xa0years. 16·0% were current smokers, 14·1% reported a history of asthma and 2·7% reported chronic obstructive pulmonary disease. Current smokers had higher T compared with never smokers (age-adjusted mean 14·5 vs 13·5xa0nmol/l, Pxa0=xa00·002) and higher E2 (65·3 vs 60·1 pmol/l, Pxa0=xa00·017). In fully adjusted analyses, T was associated with FEV1 (51xa0ml per 1 SD increase, Pxa0