EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Investigation of the structure-selectivity relationships and van't Hoff analysis of chromatographic stereoisomer separations of unusual isoxazoline-fused 2-aminocyclopentanecarboxylic acids on Cinchona alkaloid-based chiral stationary phases.
PMID 25660526
Abstract
The enantiomers of four unusual, rather rigid isoxazoline-fused 2-aminocyclopentanecarboxylic acids were directly separated on a quinine- or a quinidine-based zwitterionic ion-exchanger as chiral selector. The effects of the mobile phase composition, the structures of the analytes and temperature on the separations were investigated. Experiments were performed at constant mobile phase composition in the temperature range 10-50°C to study the effects of temperature, and thermodynamic parameters were calculated from plots of ln α versus 1/T. Some mechanistic aspects of the chiral recognition process are discussed with respect to the structures of the analytes. It was found that the enantiomer separations were in most cases predominantly enthalpy-driven, but entropically-driven separations were also observed. The sequence of elution of the enantiomers was determined in all cases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19480
Butylamine, purum, ≥99.0% (GC)
C4H11N
W313009
Butylamine, ≥99%
C4H11N
471305
Butylamine, 99.5%
C4H11N
90893
Butylamine, analytical reference material
C4H11N
D0806
Diethylamine, BioXtra
C4H11N
31730
Diethylamine, puriss. p.a., ≥99.5% (GC)
C4H11N
386456
Diethylamine, purified by redistillation, 99.5%
C4H11N
471216
Diethylamine, ≥99.5%
C4H11N
02941
Ethylamine, anhydrous, ≥99.0%
C2H7N
02946
Ethylamine, anhydrous, ≥99.5%
C2H7N
243191
Ethylamine, 97%
C2H7N
301264
Ethylamine, 97%
C2H7N
724033
Ethylamine, 99.0%
C2H7N
395064
Ethylamine solution, 2.0 M in methanol
C2H7N
395072
Ethylamine solution, 2.0 M in THF
C2H7N
471208
Ethylamine solution, 66.0-72.0% in H2O
C2H7N
31960
N,N-Diethylaniline, puriss. p.a., ≥99.5%
C10H15N
31970
N,N-Diethylaniline, purum, ≥98.0% (GC)
C10H15N
185868
N,N-Diethylaniline, ≥99%
C10H15N
240958
Propylamine, ≥99%
C3H9N
109819
Propylamine, 98%
C3H9N
53646
Propylamine, analytical standard
C3H9N
82100
Propylamine, purum, ≥99.0% (GC)
C3H9N
90781
Tributylamine, puriss. plus, ≥99.5% (GC)
C12H27N
90780
Tributylamine, puriss. p.a., ≥99.0% (GC)
C12H27N
471313
Tributylamine, ≥98.5%
C12H27N
143979
Tripropylamine, ≥98%
C9H21N
W424701
Tripropylamine, ≥98%
C9H21N