EMAIL THIS PAGE TO A FRIEND
Food chemistry
Exogenous acetaldehyde as a tool for modulating wine color and astringency during fermentation.
PMID 25660852
Abstract
Wine tannins undergo modifications during fermentation and storage that can decrease their perceived astringency and increase color stability. Acetaldehyde acts as a bridging compound to form modified tannins and polymeric pigments that are less likely to form tannin-protein complexes than unmodified tannins. Red wines are often treated with oxygen in order to yield acetaldehyde, however this approach can lead to unintended consequences due to the generation of reactive oxygen species. The present study employs exogenous acetaldehyde at relatively low and high treatment concentrations during fermentation to encourage tannin modification without promoting potentially deleterious oxidation reactions. The high acetaldehyde treatment significantly increased polymeric pigments in the wine without increasing concentrations of free and sulfite-bound acetaldehyde. Protein-tannin precipitation was also significantly decreased with the addition of exogenous acetaldehyde. These results indicate a possible treatment of wines early in their production to increase color stability and lower astringency of finished wines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1788
(±)-Catechin hydrate
C15H14O6 · xH2O
22130
(±)-Catechin hydrate, purum, ≥96.0% (HPLC)
C15H14O6 · xH2O
C1251
(+)-Catechin hydrate, ≥98% (HPLC), powder
C15H14O6 · xH2O
22110
(+)-Catechin hydrate, ≥96.0% (sum of enantiomers, HPLC)
C15H14O6 · xH2O
00070
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
C2H4O
402788
Acetaldehyde, ACS reagent, ≥99.5%
C2H4O
00071
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
C2H4O
W200336
Acetaldehyde, natural, ≥99%, FG
C2H4O
W200301
Acetaldehyde, ≥99%, meets FCC analytical specification
C2H4O
W200360
Acetaldehyde, ≥99%, meets FCC analytical specification
C2H4O
110078
Acetaldehyde, ReagentPlus®, 99%
C2H4O
W200320
Acetaldehyde, ≥99%, FCC, stabilized
C2H4O
506788
Acetaldehyde, analytical standard
C2H4O
W200379
Acetaldehyde solution, 40 wt. % in H2O
C2H4O
W200387
Acetaldehyde solution, 40 wt. % in isopropanol
C2H4O
W200344
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
C2H4O
W200328
Acetaldehyde solution, 50 wt. % in ethanol
C2H4O
719099
Acetaldehyde solution, 5 M in THF
C2H4O
W200300
Acetaldehyde solution, natural, 50 wt. % in ethanol
C2H4O
236489
Iron(III) chloride hexahydrate, ACS reagent, 97%
Cl3Fe · 6H2O
F2877
Iron(III) chloride hexahydrate, reagent grade, ≥98%, chunks
Cl3Fe · 6H2O
44944
Iron(III) chloride hexahydrate, puriss. p.a., ACS reagent, crystallized, 98.0-102% (RT)
Cl3Fe · 6H2O
31232
Iron(III) chloride hexahydrate, puriss. p.a., Reag. Ph. Eur., ≥99%
Cl3Fe · 6H2O
92816
Maleic acid, Standard for quantitative NMR, TraceCERT®
C4H4O4
63189
Maleic acid, tested according to Ph.Eur.
C4H4O4
M0375
Maleic acid, ReagentPlus®, ≥99.0% (HPLC)
C4H4O4
PHR1272
Maleic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C4H4O4
1374500
Maleic acid, United States Pharmacopeia (USP) Reference Standard
C4H4O4
PHR1472
Tartaric Acid, Pharmaceutical Secondary Standard; Certified Reference Material
C4H6O6
50926
Triethanolamine, analytical reference material
C6H15NO3