EMAIL THIS PAGE TO A FRIEND
Clinical immunology (Orlando, Fla.)
Celastrol, a Chinese herbal compound, controls autoimmune inflammation by altering the balance of pathogenic and regulatory T cells in the target organ.
PMID 25660987
Abstract
Inflammation is an integral component of autoimmune arthritis. The balance of pathogenic T helper 17 (Th17) and protective T regulatory (Treg) cells can influence disease severity, and its resetting offers an attractive approach to control autoimmunity. We determined the frequency of Th17 and Treg in the joints of rats with adjuvant arthritis (AA), a model of rheumatoid arthritis (RA). We also investigated the impact of Celastrol, a bioactive compound from the traditional Chinese medicine Celastrus that can suppress AA, on Th17/Treg balance in the joints. Celastrol treatment reduced Th17 cells but increased Treg in the joints, and it inhibited Th17 differentiation but promoted Treg differentiation in vitro by blocking the activation of pSTAT3. Furthermore, Celastrol limited the production of Th17-differentiating cytokines and chemokines (CCL3, CCL5). Thus, Celastrol suppressed arthritis in part by altering Th17/Treg ratio in inflamed joints, and it should be tested as a potential adjunct/alternative for RA therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22573F Anti-BB1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501656
Anti-CD80 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405584
Anti-CD80 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411654
Anti-CD86 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1412683
ANTI-IFNA1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410692
Anti-IFNA1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1406423
Anti-SELL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4700489 Monoclonal Anti-CD62L antibody produced in mouse, clone LT-TD180, purified immunoglobulin, buffered aqueous solution
SAB4700493 Monoclonal Anti-CD62L antibody produced in mouse, clone DREG56, purified immunoglobulin, buffered aqueous solution
SAB4700619 Monoclonal Anti-CD62L antibody produced in mouse, clone IVA94, purified immunoglobulin, buffered aqueous solution
SAB4700590 Monoclonal Anti-CD62L antibody produced in rat, clone MEL-14, purified immunoglobulin, buffered aqueous solution
WH0000941M1 Monoclonal Anti-CD80 antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1403647
Monoclonal Anti-CD80 antibody produced in mouse, clone 3E10, ascites fluid
SAB4700140 Monoclonal Anti-CD80 antibody produced in mouse, clone MEM-233, purified immunoglobulin, buffered aqueous solution
WH0000942M1 Monoclonal Anti-CD86 antibody produced in mouse, clone 3D7, purified immunoglobulin, buffered aqueous solution
SAB4700144 Monoclonal Anti-CD86 antibody produced in mouse, clone BU63, purified immunoglobulin, buffered aqueous solution
SAB4700560 Monoclonal Anti-Cd86 antibody produced in rat, clone GL-1, purified immunoglobulin, buffered aqueous solution
WH0003439M7
Monoclonal Anti-IFNA1 antibody produced in mouse, clone 3C6, purified immunoglobulin, buffered aqueous solution