EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Highly sensitive quantification of pyrethroid insecticide etofenprox in vegetables with high-performance liquid chromatography and fluorescence detection.
PMID 25662063
Abstract
This paper describes a highly sensitive analytical method using high-performance liquid chromatography and fluorescence detection (HPLC-FLD) capable of quantifying trace amounts of synthetic pyrethroid insecticide etofenprox residue in six vegetable samples: bell pepper, cucumber, eggplant, Japanese mustard spinach, spinach, and tomato. After extraction with acetonitrile, the crude sample extract was cleaned up with a solid-phase extraction cartridge. The matrix interference derived from the tested vegetable samples was evaluated. Quantification was conducted using external calibrators prepared in pure acetonitrile. The limits of quantification for etofenprox in each sample were 1.87-3.87 ng/g. Recoveries obtained by application of the proposed analytical method of vegetable samples spiked at the considerably low levels (5-100 ng/g) were 85-111% with relative standard deviations of less than 12%. The proposed method using the HPLC-FLD was applied for trace analysis of the insecticide residue in vegetable samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34094
Etofenprox, PESTANAL®, analytical standard
C25H28O3