EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Fabrication and application of zinc-zinc oxide nanosheets coating on an etched stainless steel wire as a selective solid-phase microextraction fiber.
PMID 25662065
Abstract
A novel zinc-zinc oxide (Zn-ZnO) nanosheets coating was directly fabricated on an etched stainless steel wire substrate as solid-phase microextraction (SPME) fiber via previous electrodeposition of robust Zn coating. The scanning electron micrograph of the Zn-ZnO nanosheets coated fiber exhibits a flower-like nanostructure with high surface area. The SPME performance of as-fabricated fiber was investigated for the concentration and determination of polycyclic aromatic hydrocarbons, phthalates and ultraviolet (UV) filters coupled to high performance liquid chromatography with UV detection (HPLC-UV). It was found that the Zn-ZnO nanosheets coating exhibited high extraction capability, good selectivity and rapid mass transfer for some UV filters. The main parameters affecting extraction performance were investigated and optimized. Under the optimized conditions, the calibration graphs were linear over the range of 0.1-200μgL(-1). The limits of detection of the proposed method were 0.052-0.084μgL(-1) (S/N=3). The single fiber repeatability varied from 5.18% to 7.56% and the fiber-to-fiber reproducibility ranged from 6.74% to 8.83% for the extraction of spiked water with 50μgL(-1) UV filters (n=5). The established SPME-HPLC-UV method was successfully applied to the selective concentration and sensitive determination of target UV filters from real environmental water samples with recoveries from 85.8% to 105% at the spiking level of 10μgL(-1) and 30μgL(-1). The relative standard deviations were below 9.7%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
78848
2-Ethylhexyl 4-methoxycinnamate, 98%
C18H26O3
55529
2-Ethylhexyl 4-methoxycinnamate, analytical standard
C18H26O3
W514500
2-Ethylhexyl salicylate, ≥99%
C15H22O3
52184
2-Ethylhexyl salicylate, analytical standard
C15H22O3
437379
2-Ethylhexyl salicylate, 99%
C15H22O3
01479
2-Hydroxy-4-methoxybenzophenone, certified reference material, TraceCERT®
C14H12O3
H36206
2-Hydroxy-4-methoxybenzophenone, 98%
C14H12O3
07671
Anthracene, certified reference material, TraceCERT®
C14H10
31581
Anthracene, analytical standard
C14H10
48567
Anthracene, analytical standard
C14H10
10580
Anthracene, suitable for scintillation, ≥99.0% (GC)
C14H10
331481
Anthracene, zone-refined, ≥99%
C14H10
141062
Anthracene, ReagentPlus®, 99%
C14H10
A89200
Anthracene, reagent grade, 97%
C14H10
694959
Anthracene, sublimed grade, ≥99%
C14H10
ERA-010
Anthracene, vial of 250 mg, analytical standard
C14H10
40076
Anthracene solution, certified reference material, 1000 μg/mL in acetone
C14H10
48647
Anthracene solution, certified reference material, 200 μg/mL in methanol
C14H10
67261
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®
C24H38O4
36735
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
C24H38O4
80030
Bis(2-ethylhexyl) phthalate, Selectophore
C24H38O4
89353
Density Standard 1191 kg/m3, H&D Fitzgerald Ltd. Quality
C10H10O4
88173
Di-n-octyl phthalate, certified reference material, TraceCERT®
C24H38O4
31301
Di-n-octyl phthalate, analytical standard
C24H38O4
80153
Di-n-octyl phthalate, ≥98.0% (GC)
C24H38O4
48560-U
Di-n-octyl phthalate, analytical standard
C24H38O4
18281
Dibutyl phthalate, certified reference material, TraceCERT®
C16H22O4
D1010000
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
C16H22O4
80100
Dibutyl phthalate, Selectophore
C16H22O4
36736
Dibutyl phthalate, PESTANAL®, analytical standard
C16H22O4