EMAIL THIS PAGE TO A FRIEND
The Journal of pathology
BCR-JAK2 drives a myeloproliferative neoplasm in transplanted mice.
PMID 25664618
Abstract
BCR-JAK2 is an infrequent gene fusion found in chronic/acute, myeloid/lymphoid Philadelphia chromosome-negative leukaemia. In this study, we demonstrated that in vivo expression of BCR-JAK2 in mice induces neoplasia, with fatal consequences. Transplantation of BCR-JAK2 bone marrow progenitors promoted splenomegaly, with megakaryocyte infiltration and elevated leukocytosis of myeloid origin. Analysis of peripheral blood revealed the presence of immature myeloid cells, platelet aggregates and ineffective erythropoiesis. A possible molecular mechanism for these observations involved inhibition of apoptosis by deregulated expression of the anti-apoptotic mediator Bcl-xL and the serine/threonine kinase Pim1. Together, these data provide a suitable in vivo molecular mechanism for leukaemia induction by BCR-JAK2 that validates the use of this model as a relevant preclinical tool for the design of new targeted therapies in Philadelphia chromosome-negative leukaemia involving BCR-JAK2-driven activation of the JAK2 pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1305652
ANTI-CD11B(N-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2500547
Anti-ITGAM antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4300394
Anti-JAK2 (Ab-1007) antibody produced in rabbit, affinity isolated antibody
SAB4300587
Anti-JAK2 (Ab-570) antibody produced in rabbit, affinity isolated antibody
SAB2501574 Anti-JAK2 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4501599
Anti-JAK2 antibody produced in rabbit, affinity isolated antibody
SAB4501601
Anti-JAK2 antibody produced in rabbit, affinity isolated antibody
SAB4501600
Anti-JAK2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501602
Anti-JAK2, C-Terminal antibody produced in rabbit, affinity isolated antibody
AMAB90911
Monoclonal Anti-ITGAM antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1719, purified immunoglobulin, buffered aqueous glycerol solution
SAB1402826 Monoclonal Anti-ITGAM antibody produced in mouse, clone 4E10, purified immunoglobulin, buffered aqueous solution
SAB4200483
Monoclonal Anti-JAK2 antibody produced in mouse, ~1.0 mg/mL, clone 10.1.5, purified immunoglobulin
SAB5300075
Monoclonal Anti-JAK2 antibody produced in mouse, clone 1C1, ascites fluid