EMAIL THIS PAGE TO A FRIEND
European journal of clinical pharmacology
Pharmacokinetic interaction between HCV protease inhibitor boceprevir and methadone or buprenorphine in subjects on stable maintenance therapy.
PMID 25666027
Abstract
Intravenous opioid use is a common route of hepatitis C virus (HCV) infection; consequently, the prevalence of HCV is high among patients on methadone or buprenorphine/naloxone. The authors evaluated the pharmacokinetic interaction of boceprevir with methadone or buprenorphine/naloxone in patients on stable maintenance therapy. This was a two-center, open-label, fixed-sequence study in 21 adult volunteers on stable maintenance therapy. Oral methadone (20-150 mg once daily) or sublingual buprenorphine/naloxone (8/2-24/6 mg once daily) was administered alone or in combination with boceprevir (800 mg every 8 h) on days 2-7. Pharmacokinetic sampling occurred before and up to 24 h after the dose on days 1 and 7. Coadministration of boceprevir reduced the area under the concentration-time curve during a dosing interval τ (AUC τ ) and maximum observed plasma (or serum) concentration (C max) of R-methadone (geometric mean ratios (GMRs) [90 % confidence intervals (CIs)], 0.85 [0.74, 0.96] and 0.90 [0.71, 1.13]) and S-methadone (GMRs [90 % CIs], 0.78 [0.66, 0.93] and 0.83 [0.64, 1.09]). Boceprevir increased the AUC τ and C max of buprenorphine (GMRs [90 % CIs], 1.19 [0.91, 1.58] and 1.18 [0.93, 1.50]) and naloxone (GMRs [90 % CIs], 1.33 [0.90, 1.93] and 1.09 [0.79, 1.51]). Boceprevir exposure upon methadone or buprenorphine/naloxone coadministration was not clinically different from historical controls and there was no evidence of opioid withdrawal or excess. There was no clinically meaningful impact of boceprevir on methadone or buprenorphine pharmacokinetics, suggesting that methadone/buprenorphine dose adjustments are not required upon coadministration with boceprevir. Individual patients may differ in their clinical experience and clinicians should maintain vigilance when coadministering these medications.