EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Molecular characterization of the boron adducts of the proteasome inhibitor bortezomib with epigallocatechin-3-gallate and related polyphenols.
PMID 25669488
Abstract
The green tea polyphenol epigallocatechin-3-gallate (EGCG) was reported to effectively antagonize the ability of Bortezomib (BZM) to induce apoptosis in cancer cells. This interaction was attributed to the formation of a covalent adduct between a phenolic moiety of EGCG with the boronic acid group of Bortezomib. However, the structural details of this boron adduct and the molecular factors that contribute to its formation and its ability to inhibit Bortezomib's activity remain unclear. This paper describes the use of NMR spectroscopy and cell assays to characterize the structures and properties of the boron adducts of EGCG and related polyphenols. The observed boron adducts included both boronate and borate derivatives, and their structural characteristics were correlated with cell-based evaluation of the ability of EGCG and other phenols to antagonize the anticancer activity of Bortezomib. The enhanced stability of the BZM/EGCG adduct was attributed to electronic and steric reasons, and a newly identified intramolecular interaction of the boron atom of BZM with the adjacent amide bond. The reported approach provides a useful method for determining the potential ability of polyphenols to form undesired adducts with boron-based drugs and interfere with their actions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1753
(−)-Epicatechin, ≥90% (HPLC)
C15H14O6
68097
(−)-Epicatechin, analytical standard
C15H14O6
E4018
(−)-Epicatechin, ≥98% (HPLC), from green tea
C15H14O6
50299
(−)-Epigallocatechin gallate, ≥97.0% (HPLC)
C22H18O11
93894
(−)-Epigallocatechin gallate, analytical standard
C22H18O11
56753
2-Phenoxyethanol, analytical standard
C8H10O2
77699
2-Phenoxyethanol, ≥99%
C8H10O2
10832
2-Phenoxyethanol, tested according to Ph.Eur.
C8H10O2
PHR1121
2-Phenoxyethanol, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10O2
N7126
4-Nitrocatechol, Sigma Grade
C6H5NO4
N15553
4-Nitrocatechol, 97%
C6H5NO4
03940590
Epicatechin, primary pharmaceutical reference standard
C15H14O6
03970590
Epigallocatechin gallate, primary pharmaceutical reference standard
C22H18O11
PHR1333
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
C22H18O11
Y0000012
Hymecromone impurity A, European Pharmacopoeia (EP) Reference Standard
C6H6O2
C9510
Pyrocatechol, ≥99%
C6H6O2
C3561
Pyrocatechol, BioReagent, plant cell culture tested, ≥99%, powder
C6H6O2
P0381
Pyrogallol, ≥98% (HPLC)
C6H6O3
06931
Pyrogallol, analytical standard
C6H6O3
16040
Pyrogallol, ACS reagent, ≥99%
C6H6O3
254002
Pyrogallol, ACS reagent
C6H6O3
53363
Resorcinol, certified reference material, TraceCERT®
C6H6O2
R5645
Resorcinol, BioXtra, ≥99%
C6H6O2
398047
Resorcinol, ACS reagent, ≥99.0%
C6H6O2
307521
Resorcinol, ReagentPlus®, 99%
C6H6O2
R406
Resorcinol, reagent grade, 98%
C6H6O2
16101
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
C6H6O2
83601
Resorcinol, tested according to Ph.Eur.
C6H6O2
W358908
Resorcinol, ≥98%, FG
C6H6O2
Y0000767
Resorcinol, European Pharmacopoeia (EP) Reference Standard
C6H6O2