EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
Tenellin acts as an iron chelator to prevent iron-generated reactive oxygen species toxicity in the entomopathogenic fungus Beauveria bassiana.
PMID 25670702
Abstract
Iron is an essential element for life. However, the iron overload can be toxic. Here, we investigated the significant increase of tenellin and iron-tenellin complex production in ferricrocin-deficient mutants of Beauveria bassiana. Our chemical analysis indicated that the ferricrocin-deficient mutants T1, T3 and T5 nearly abolished ferricrocin production. In turn, these mutants had significant accumulation of iron-tenellin complex in their mycelia at 247-289 mg g(-1) cell dry weight under iron-replete condition. Both tenellin and iron-tenellin complex were not detected in the wild-type under such condition. Mass analysis of the mutants' crude extracts demonstrated that tenellin formed a 3:1 complex with iron in the absence of ferricrocin. The unexpected link between ferricrocin and tenellin biosynthesis in ferricrocin-deficient mutants could be a survival strategy during iron-mediated oxidative stress.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N10802
2-Nitrobenzaldehyde, 98%
C7H5NO3
SAB2502094
Anti-CHRM2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405619
Anti-CHRM2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA029795
Anti-CHRM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500245
Anti-CHRM2 antibody produced in rabbit, affinity isolated antibody
SAB3700280 Anti-goat IgG (H+L) antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB3700328 Anti-Goat IgG (H+L), F(ab) fragment- antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB3701079 Anti-Mouse IgG (H+L) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701163 Anti-Mouse IgG (H+L), F(ab) fragment antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3700999 Anti-Mouse IgG F(ab′)2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701000 Anti-Mouse IgG F(ab′)2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
M9693 Anti-Muscarinic Acetylcholine Receptor M2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA034660
Anti-NOTO antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105242
Anti-NOTO, (N-terminal) antibody produced in rabbit, affinity isolated antibody
M7753
Maltoheptaose, ≥60% (HPLC)
C42H72O36
WH0001129M1
Monoclonal Anti-CHRM2 antibody produced in mouse, clone 4B5, purified immunoglobulin, buffered aqueous solution