EMAIL THIS PAGE TO A FRIEND
Journal of biomedical optics
Multidimensional visualization of healthy and sensitized rabbit knee tissues by means of confocal microscopy.
PMID 25672969
Abstract
This study combines several fluorescence detection methods to distinguish structural features of the synovium and cartilage tissues and to visualize the localization of endogenous porphyrins in the sensitized tissues. Specimens of synovium and cartilage tissues obtained from rabbits with antigen-induced monoarthritis after intra-articular 5-aminolevulinic acid methyl ester injection and those from healthy rabbits were investigated ex vivo by means of fluorescence spectroscopy, fluorescence intensity, and lifetime microscopy. The presence of endogenous porphyrins was confirmed with the fluorescence spectra measured on sliced sensitized specimens. Application of the lifetime-gating method on fast fluorescence lifetime imaging microscopy images, allowed separate visualization of tissue structures possessing different average lifetimes. The presence of the structures has been validated by histopathological imaging based on conventional rapid hematoxylin–eosin staining of the specimens. The fluorescence lifetime of endogenous protoporphyrin IX has been assessed and employed for visualization of sensitized tissues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5575
Methyl δ-aminolevulinate hydrochloride, ≥98% (TLC)
C6H11NO3 · HCl