EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Modulation of microglial process convergence toward neuronal dendrites by extracellular calcium.
PMID 25673836
Abstract
Extracellular calcium concentrations in the brain fluctuate during neuronal activities and may affect the behavior of brain cells. Microglia are highly dynamic immune cells of the brain. However, the effects of extracellular calcium concentrations on microglial dynamics have not been investigated. Here, we addressed this question in mouse brain slices and in vivo using two-photon microscopy. We serendipitously found that extracellular calcium reduction induced microglial processes to converge at distinct sites, a phenomenon we termed microglial process convergence (MPCs). Our studies revealed that MPCs target neuronal dendrites independent of neuronal action potential firing and is mediated by ATP release and microglial P2Y12 receptors. These results indicate that microglia monitor and interact with neurons during conditions of cerebral calcium reduction in the normal and diseased brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7635
Sulforhodamine 101
C31H30N2O7S2