EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Change of surface structure upon foam film formation.
PMID 25677129
Abstract
The charge distribution and coverage with surfactant molecules at foam film surfaces plays an important role in determining foam film structure and stability. This work uses the concentration depth profiling technique neutral impact collision ion scattering spectroscopy to experimentally observe the charge distribution in a foam film for the first time. The charge distribution at the surface of a foam film and the surface of the corresponding bulk liquid were measured for a cationic surfactant solution and the surface excess as well as the electric potential were determined. Describing the internal pressure of foam films by using the electrochemical potential is introduced as a new concept. The foam film can be seen to have a more negative surface charge compared to the bulk liquid surface due to re-arranging of the surfactant molecules. It is discussed how the change in surface excess and electric potential change the electrochemical potential and the stability of the foam film.