EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
b-Series gangliosides crucially regulate leptin secretion in adipose tissues.
PMID 25677621
Abstract
Gangliosides are widely involved in the regulation of cells and organs. However, little is known about their roles in leptin secretion from adipose tissues. Genetic deletion of b-series gangliosides resulted in the marked reduction of serum leptin. Expression analysis of leptin revealed that leptin accumulated in the adipose tissues of GD3 synthase-knockout (GD3S KO) mice. Analysis of primary cultured stromal vascular fractions (SVF) derived from GD3S KO mice revealed that leptin secretion was reduced, although leptin amounts in cells were increased compared with those of wild type. Interestingly, addition of b-series gangliosides to the culture medium of differentiated SVF resulted in the restoration of leptin secretion. Results of methyl-β-cyclodextrin treatment of differentiated 3T3-L1 cells as well as immunocytostaining of leptin and caveolin-1 suggested that b-series gangliosides regulate the leptin secretion from adipose tissues in lipid rafts.