EMAIL THIS PAGE TO A FRIEND
Biological trace element research
Dyshomeostasis of Serum Oxidant/Antioxidant Status and Copper, Zinc, and Selenium Levels in Elderly Physically Disabled Persons: an AHAP-Based Study.
PMID 25677848
Abstract
The percentage of elderly persons is rapidly growing. Physical disability is one of the main age-related diseases which affect life quality. There are some studies that suggest the oxidative stress and trace elements are involved in physical disability in elderly persons, but the results are inconclusive. Therefore, the aim of this study was to investigate the status of aforementioned parameters in elderly physically disabled patients vs. healthy ones. According to the Katz questionnaire form, 44 subjects with physical disability and 66 age-gender-matched healthy subjects were selected from Amirkola Health and Aging Project (AHAP). The results indicated that patient group had lower serum Zn, Se, and total antioxidant levels than the control group (p < 0.001), whereas serum total oxidant level and Cu to Zn ratio (CZr) were higher in control group than in healthy one (p < 0.001). A positive correlation was found between Zn, Se, total antioxidant, and bone mineral density of femur (BMD.F) with activities of daily living (ADL) score (p < 0.01); meanwhile, a negative correlation between CZr and total oxidant with ADL score was observed (p < 0.01). Serum total oxidant level and CZr index had the highest area under the curve in receiver operating characteristic (ROC) analysis among the included parameters for discrimination of physically disabled patients than the normal ones. Decrease in serum Zn and Se levels, low BMD, and increase in CZr and oxidative stress were observed in physically disabled patients. It seems that CZr is more reliable parameter than the others to discriminate the physically disabled patients than the healthy persons.