EMAIL THIS PAGE TO A FRIEND
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology
Epidermal growth factor is a potential biomarker for poor cetuximab response in tongue cancer cells.
PMID 25677871
Abstract
Head and neck squamous cell carcinoma is frequently associated with aberrant epidermal growth factor receptor (EGFR) signaling, which contributes to tumor growth. Here, the functional importance of EGFR ligands in relation to proliferation and sensitivity to the EGFR-targeted therapy cetuximab was investigated in three tongue cancer cell lines. The influence of epidermal growth factor (EGF), amphiregulin (AR), and epiregulin (EPR) on tumor cell proliferation and cetuximab response was evaluated by the addition of recombinant human (rh) proteins or by siRNA-mediated downregulation of the endogenous ligand production. The expression, activation and cellular distribution of EGFR were assessed by Western blot analysis and immunocytochemistry. EGF downregulation suppressed the proliferation of all investigated tumor cell lines, whereas the response to an increased level of EGF differed between EGFR-overexpressing and EGFR-non-overexpressing cell lines. Furthermore, tumor cells consistently displayed increased cetuximab resistance upon the addition of rhEGF, whereas EGF silencing was associated with an improved cetuximab response. The data regarding AR and EPR were inconclusive. Our data suggest that the amount of EGF is a determinant of the tumor cell proliferation rate and the response to cetuximab treatment in tongue cancer. Thus, EGF is a potential predictive biomarker of poor cetuximab response and a possible treatment target.