EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
LDL-R promoting activity of peptides derived from human PCSK9 catalytic domain (153-421): design, synthesis and biochemical evaluation.
PMID 25679794
Abstract
High level of Low Density Lipoprotein-Cholesterol (LDL-C) in circulation in the blood is associated with an elevated risk of cardiovascular disease (CVD) and stroke. Currently the statin drugs which inhibit the enzyme HMG-CoA reductase responsible for cholesterol synthesis in the liver are very effective in lowering LDL-cholesterol. However these drugs are often associated with serious side effects particularly for ∼10-12% of cases. Therefore there is a need to develop non-statin based cholesterol reducing agents. Recently it was revealed that the secreted Proprotein Convertase Subtilisin Kexin 9 (PCSK9) binds with LDL-receptor (LDL-R) causing its degradation in the lysosome with the result of LDL-C accumulating in the blood. Thus PCSK9 has become an alternative target for development of non-statin cholesterol reducing agents. It is established that the catalytic domain of PCSK9 (aa153-421) and the EGF-A domain of LDL-R (aa314-355) are involved in the above bind leading to the reduction of LDL-R level and accumulation of LDL-C. The major goal of this study is to identify peptide/s from the catalytic domain of hPCSK9 that can block the binding of hPCSK9 and LDL-R and therefore can reduce LDL-R degradation leading to the clearance of LDL-C from the plasma. Using 51 synthetic linear peptides (P1-P51) of 15aa long with 10 amino acids overlapping sequences spanning the entire catalytic segment of hPCSK9 (aa153-421), we identified two domains of hPCSK9 namely (aa323-358) and (aa365-384) that exhibited strong binding affinity towards synthetic EGF-A peptide. The results were based on mass spectrometry, fluorescence spectroscopy and native gel electrophoresis. Thus peptides containing the above segments in part (P35-P39 and P42-P47) exhibited LDL-R promoting activity when added exogenously to culture medium of growing human hepatic cells like HepG2 and HuH7. The effects were particularly significant with peptides P36, P37, P46 and P47. Interestingly, the first two peptides are present within the disulphide loop Cys(323)-Cys(358) and contain the key gain of function mutation D(374)/Y site while the last two peptides contain another disulphide bridge loop Cys(375)-Cys(378) and the second most potent gain of function mutation R(357)/H. Further studies revealed that S-S bridged cyclic loop peptide hPCSK9(365-384) exhibited the highest (∼3.5-fold) LDL-R promoting activity in both HepG2 and HuH7 when applied at 5xa0μM concentration level. This effect is completely abrogated when one of the Cys residues is substituted by Ala thereby preventing any S-S bond formation. This suggested its critical role in the bioactivity. It is proposed that LDL-R promoting activity of this and other selected PCSK9 catalytic peptides such as P36, P37, P46 and P47 are most likely mediated via intervention of PCSK9:LDL-R complex formation. Our findings may find useful application in future development of small molecule PCSK9 inhibitors for intervention of hypercholesterolemia and associated cardiovascular disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
39319
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
C7H6O4
78069
2,5-Dihydroxybenzoic acid, analytical standard
C7H6O4
242381
Benzoic acid, ACS reagent, ≥99.5%
C7H6O2
33045
Benzoic acid, for calorimetrical determination (approx. 26460 J/g)
C7H6O2
06185
Benzoic acid, Standard for quantitative NMR, TraceCERT®
C7H6O2
1055002
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
C7H6O2
33047
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
C7H6O2
18102
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
C7H6O2
12356
Benzoic acid, tested according to Ph.Eur.
C7H6O2
PHR1050
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O2
12353
Benzoic acid, certified reference material for titrimetry, certified by BAM according to ISO 17025, ≥99.5%
C7H6O2
47849
Benzoic acid, analytical standard
C7H6O2
427608
Benzoic acid, purified by sublimation, ≥99%
C7H6O2
109479
Benzoic acid, ReagentPlus®, 99%
C7H6O2
W213101
Benzoic acid, ≥99.5%, FCC, FG
C7H6O2
W213128
Benzoic acid, natural, ≥99.5%, FCC, FG
C7H6O2
Y0001470
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
C7H6O2
491861
Benzoic acid-12C7, 99.9 atom % 12C
12C7H6O2
76170
Melting point standard 121-123°C, analytical standard
C7H6O2
73983
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C7H6O2
295647
Phosphine, electronic grade, ≥99.9995%
H3P
04004
Phosphorus, red, ≥97.0%
P
343242
Phosphorus, red, ≥99.99% trace metals basis
P
209678
Phosphorus, red, powder, 99% trace metals basis
P
49508
Sinapic acid, matrix substance for MALDI-MS, ≥99.5%, Ultra pure
C11H12O5
85429
Sinapic acid, matrix substance for MALDI-MS, ≥99.0% (T)
C11H12O5
D7927
Sinapic acid, ≥98%, powder
C11H12O5
8S61336 SS Benzoic Acid, 2000 μg/mL in dichloromethane, analytical standard
C7H6O2
93878
trans-Sinapic acid, analytical standard
C11H12O5