EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
CD73 plays a protective role in collagen-induced arthritis.
PMID 25681339
Abstract
Rheumatoid arthritis (RA) is a chronic autoimmune disease with significant morbidity and mortality. Recent studies suggest that modulation of adenosine signaling, a potent immunosuppressive pathway, is a promising approach for treatment of RA. Extracellular adenosine can come from two sources: transport of intracellular adenosine and hydrolysis of extracellular adenine nucleotides by CD73. In this study, we investigated the susceptibility of CD73-deficient C57BL/6 mice to collagen-induced arthritis (CIA), a well-established mouse model of RA. Our data demonstrated that CD73-deficient mice are significantly more susceptible to CIA than wild-type mice. CD73 deficiency resulted in an increased production of proinflammatory cytokines in the joints, increased Th1 T cell responses, and increased joint destruction. Surprisingly, this was accompanied by delayed anticollagen IgG responses, suggesting defective isotype class switching in CD73-deficient mice. Using bone marrow chimera mice, we demonstrated that CD73 expression on nonhematopoietic cells, but not on hematopoietic cells, was important for protection from CIA. We further demonstrated that administration of a selective A2A adenosine receptor agonist to CD73-deficient mice resulted in arthritis incidence similar to wild-type mice in support of a protective role for A2A signaling. Taken together, our study identifies CD73 as an important regulator of CIA in mice. It also strengthens the notion that CD73-generated adenosine by nonhematopoietic cells plays a protective role in RA and suggests that strategies able to enhance CD73 activity or expression levels may be a valid therapeutic option.