EMAIL THIS PAGE TO A FRIEND
Forensic science international. Genetics
SNP-microarrays can accurately identify the presence of an individual in complex forensic DNA mixtures.
PMID 25682311
Abstract
Common forensic and mass disaster scenarios present DNA evidence that comprises a mixture of several contributors. Identifying the presence of an individual in such mixtures has proven difficult. In the current study, we evaluate the practical usefulness of currently available "off-the-shelf" SNP microarrays for such purposes. We found that a set of 3000 SNPs specifically selected for this purpose can accurately identify the presence of an individual in complex DNA mixtures of various compositions. For example, individuals contributing as little as 5% to a complex DNA mixture can be robustly identified even if the starting DNA amount was as little as 5.0ng and had undergone whole-genome amplification (WGA) prior to SNP analysis. The work presented in this study represents proof-of-principle that our previously proposed approach, can work with real "forensic-type" samples. Furthermore, in the absence of a low-density focused forensic SNP microarray, the use of standard, currently available high-density SNP microarrays can be similarly used and even increase statistical power due to the larger amount of available information.