EMAIL THIS PAGE TO A FRIEND
Cancer letters
Cofilin-phosphatase slingshot-1L (SSH1L) is over-expressed in pancreatic cancer (PC) and contributes to tumor cell migration.
PMID 25684665
Abstract
Slingshot-1L (SSH1L), a cofilin-phosphatase, plays a role in actin dynamics and cell migration by reactivating cofilin-1. However, the expression of SSH1L in malignant diseases is poorly understood. The overexpression of SSH1L in cancerous tissue compared to the matched surrounding non-cancerous tissues from patients with late stages (III-IV) of PC was detected in 90% (9/10) of cases by western blotting. The expression of SSH1L was shown to be upregulated in tumor cells from 10.7% (11/102) of patients with pancreatic cancer (PC) by immunohistochemistry (IHC). The positive rate of SSH1L in patients with PC at stage VI (TNM) categorized as grade 3 was of 50% (2/4) and 15% (6/40), respectively. Moreover, SSH1L expression was shown to be up-regulated in the PC cell lines (KLM1, PANC-1 and MIAPaCa-2) with high metastatic potential. Loss of SSH1L expression was associated with an increase in the phosphorylation of cofilin-1 at serine-3 and further inhibited cell migration (but not proliferation) in KLM1, PANC-1 and MIAPaCa-2. Actin polymerization inhibitor cytochalasin-D was sufficient to abrogate cell migration of PC without changing SSH1L expression. These results reveal that SSH1L is upregulated in a subset of PCs and that the SSH1L/cofilin-1 signal pathway is associated positively in PC with cell migration. Our study may thus provide potential targets to prevent and/or treat PC invasion and metastasis in patients with SSH1L-positive PC.