EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Channel characteristics of VDAC-3 from Arabidopsis thaliana.
PMID 25686492
Abstract
Four different isoforms of the Voltage-Dependent Anion Channel (VDAC) have been identified in Arabidopsis plant cells. The electrophysiological characteristics of several VDAC channels from animal as well as plant cells are well documented, but those of this model plant are unknown. One isoform, AtVDAC-3 was obtained either directly by cell-free synthesis or produced in Escherichia coli, as inclusion bodies, and re-natured. An electrophysiological study of the purified proteins in planar lipid bilayers showed that both methods yielded proteins with similar channel activity. The characteristics of AtVDAC-3 are that of a bona fide VDAC-like channel.