EMAIL THIS PAGE TO A FRIEND
Oncotarget
RACK1 is a candidate gene associated with the prognosis of patients with early stage non-small cell lung cancer.
PMID 25686824
Abstract
This study was conducted to identify genetic polymorphisms associated with the prognosis of patients with early stage NSCLC. We genotyped 1,969 potentially functional single nucleotide polymorphisms (SNPs) of 1,151 genes involved in carcinogenesis in 166 NSCLC patients who underwent curative surgery, using the Affymetrix custom-made GeneChip. A replication study was performed in an independent cohort of 626 patients. Fifty six SNPs which were associated with both overall survival (OS) and disease-free survival (DFS) with log-rank P values < 0.05 in discovery set were selected for validation. Among those, five SNPs (RACK1 rs1279736C>A and rs3756585T>G, C3 rs2287845T>C, PCAF rs17006625A>G, and PCM1 rs17691523C>G) were found to be significantly associated with survival in the same direction as the discovery set. In combined analysis, the rs1279736C>A and rs3756585T>G were most significantly associated with OS and DFS in multivariate analysis (P for OS = 4 × 10⁻⁵ and 7 × 10⁻⁵, respectively; and P for DFS = 0.003, both; under codominant model). In vitro promoter assay and electrophoretic mobility shift assay revealed that the rs3756585 T-to-G change increased promoter activity and transcription factor binding of RACK1. We identified five SNPs, especially RACK1 rs3756585T>G, as markers for prognosis of patients with surgically resected NSCLC.