EMAIL THIS PAGE TO A FRIEND
Parasitology research
Inducible nitric oxide synthase response and associated cytokine gene expression in the spleen of mice infected with Clonorchis sinensis.
PMID 25687522
Abstract
Clonorchis sinensis is a food-borne parasite that induces a permanent increase of nitrosation in the body upon infection. The spleen is an important secondary lymphoid organ for the regulation of immune responses locally and in the whole body. However, the functions and mechanisms of the spleen in nitric oxide (NO) responses after C. sinensis infection remain unknown. In this study, BALB/c mice were infected with 20, 40, and 80 C. sinensis metacercariae to simulate mild, moderate, and severe infections, respectively. We examined the expression of inducible nitric oxide synthase (iNOS) in the spleen and the relevant cytokine transcription in splenocytes from the mice infected with different amounts of metacercariae. The iNOS of the mice infected with 80 metacercariae was expressed in the spleen as early as 10 days post-infection (dpi) and gradually increased until 90 dpi. The iNOS expression in the mice infected with 40 metacercariae was detected only at 45 and 90 dpi, but not in the mice infected with 20 metacercariae. The level of interferon (IFN)-γ messenger RNA (mRNA) transcription in splenocytes significantly increased at 10 and 20 dpi (P < 0.05) in response to mild/moderate infection but gradually decreased to normal levels after 45 dpi. The level of IL-12p35 mRNA transcription did not change at 10 and 20 dpi but significantly decreased after 45 dpi under moderate/severe infection (P < 0.05/0.01/0.001). The level of IL-18 mRNA transcription significantly increased at 10 dpi (P < 0.05/0.01) but significantly decreased after 20 dpi (P < 0.05/0.01/0.001). These results suggest that spleen is an important organ for iNOS/NO responses, which correspond to the severity of C. sinensis infection, but cannot be attributed to the expression of the Th1 cytokines.