EMAIL THIS PAGE TO A FRIEND
Cancer chemotherapy and pharmacology
A pharmacokinetic binding model for bevacizumab and VEGF165 in colorectal cancer patients.
PMID 25687989
Abstract
To characterize the population pharmacokinetics of bevacizumab, its binding properties to VEGF165 and the effect of demographic data and VEGF-A polymorphisms on the interplay between bevacizumab serum pharmacokinetics and VEGF165 serum concentrations in patients with colorectal cancer stage IV. Bevacizumab and VEGF165 data were collected from 19 adult patients with metastatic colorectal cancer enrolled in an observational clinical study. Bevacizumab was administered with one of the following combinations: 5-FU/Leucovorin/Irinotecan, 5-FU/Leucovorin/Oxaliplatin, Capecitabine/Irinotecan at doses ranging from 5 to 10xa0mg/kg every 2 or 3xa0weeks. Data analysis was performed using nonlinear mixed-effects modeling implemented in NONMEM 7.3. A target-mediated drug disposition model adequately described bevacizumab concentration changes over time and its binding characteristics to VEGF165. The estimated clearance of bevacizumab was 0.18xa0L/day, the free VEGF165 levels at baseline were 212xa0ng/L, and the elimination rate constant of free VEGF165 was 0.401xa0day(-1). Body weight was allometrically included in all PK parameters. The final model adequately described the pre- and post-dose concentrations of total bevacizumab and free VEGF165 in patients with colorectal cancer. Model parameters were consistent with those previously reported for patients with solid tumors. Correlations between the binding affinity of bevacizumab and the VEGF-2578C/A and VEGF-634G/C polymorphisms were noticed.