EMAIL THIS PAGE TO A FRIEND
Oncotarget
Gold nano-particles (AuNPs) carrying anti-EBV-miR-BART7-3p inhibit growth of EBV-positive nasopharyngeal carcinoma.
PMID 25691053
Abstract
Epstein-Barr virus (EBV) infection is a major etiological factor for nasopharyngeal carcinoma (NPC). Several EBV-encoded BART miRNAs have been associated with viral latency, immune escape, cell survival, cell proliferation and apoptosis. Here, we report that EBV-miR-BART7-3p, an EBV-encoded BART miRNA highly expressed in NPC, was correlated with cell-cycle progression in vitro and increased tumor formation in vivo. This viral miRNA stimulated the PTEN/PI3K/Akt pathway and induced c-Myc and c-Jun. Knockdown of PTEN mimicked EBV-miR-BART7-3p-induced tumorigenic phenotype. Based on these results, we conducted a therapeutic experiment by using gold nano-particles (AuNPs) carrying anti-EBV-miR-BART7-3p. Silencing of EBV-miR-BART7-3p reduced tumor growth in animal model. We conclude that EBV-miR-BART7-3p favors carcinogenesis, representing a potential target for miRNA-based therapy.