EMAIL THIS PAGE TO A FRIEND
Journal of medicinal food
Protective Effects of Quercetin Against HgCl₂-Induced Nephrotoxicity in Sprague-Dawley Rats.
PMID 25692400
Abstract
Mercury is a well-known environmental pollutant that can cause nephropathic diseases, including acute kidney injury (AKI). Although quercetin (QC), a natural flavonoid, has been reported to have medicinal properties, its potential protective effects against mercury-induced AKI have not been evaluated. In this study, the protective effect of QC against mercury-induced AKI was investigated using biochemical parameters, new protein-based urinary biomarkers, and a histopathological approach. A 250 mg/kg dose of QC was administered orally to Sprague-Dawley male rats for 3 days before administration of mercury chloride (HgCl2). All animals were sacrificed at 24 h after HgCl2 treatment, and biomarkers associated with nephrotoxicity were measured. Our data showed that QC absolutely prevented HgCl2-induced AKI, as indicated by biochemical parameters such as blood urea nitrogen (BUN) and serum creatinine (sCr). In particular, QC markedly decreased the accumulation of Hg in the kidney. Urinary excretion of protein-based biomarkers, including clusterin, kidney injury molecule-1 (KIM-1), neutrophil gelatinase-associated lipocalin (NGAL), monocyte chemoattractant protein-1 (MCP-1), tissue inhibitor of metalloproteinases 1 (TIMP-1), and vascular endothelial growth factor (VEGF) in response to HgCl2 administration were significantly decreased by QC pretreatment relative to that in the HgCl2-treated group. Furthermore, urinary excretion of metallothionein and Hg were significantly elevated by QC pretreatment. Histopathological examination indicated that QC protected against HgCl2-induced proximal tubular damage in the kidney. A TUNEL assay indicated that QC pretreatment significantly reduced apoptotic cell death in the kidney. The administration of QC provided significant protective effects against mercury-induced AKI.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1407321
Anti-HAVCR1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA007173
Anti-HAVCR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101016
Anti-HAVCR1 antibody produced in rabbit, affinity isolated antibody
SAB3700699 Anti-Sheep IgG (Fc specific), highly cross adsorbed antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3700728 Anti-Sheep IgG (H+L), F(ab′)2 fragment antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3700744 Anti-Sheep IgG (H+L), F(ab) fragment antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3700685 Anti-Sheep IgG F(ab′)2, highly cross adsorbed antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500252
Anti-TIM-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
203777
Mercury(II) chloride, 99.999% trace metals basis
Cl2Hg
449202
Mercury(II) chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Cl2Hg
429724
Mercury(II) chloride, anhydrous, powder, 99.998% trace metals basis
Cl2Hg
413445
Mercury(II) chloride, ≥98%
Cl2Hg
WH0026762M1
Monoclonal Anti-HAVCR1 antibody produced in mouse, clone 1E5, purified immunoglobulin, buffered aqueous solution
74988
Octylamine, ≥99.5%
C8H19N
O5802
Octylamine, 99%
C8H19N