EMAIL THIS PAGE TO A FRIEND
Journal of labelled compounds & radiopharmaceuticals
Synthesis of deuterium-labelled isotopomer of deferasirox.
PMID 25693166
Abstract
A d4 -labeled isotopomer of deferasirox was synthesized as internal standard for use in a LC/mass spectroscopy (MS)/MS method developed for the simultaneous quantitative determination of deferasirox in human serum. d4 -deferasirox was synthesized from d8 -toluene.