EMAIL THIS PAGE TO A FRIEND
Preventing chronic disease
Trends in major risk factors for cardiovascular disease among adults in the Mississippi Delta region, Mississippi Behavioral Risk Factor Surveillance System, 2001-2010.
PMID 25695259
Abstract
The prevalences of major modifiable risk factors for cardiovascular disease (CVD) are disproportionately high in the 18-county Mississippi Delta region, and many of these risk factors disproportionately affect blacks. Temporal trends in the prevalence of CVD risk factors in the Mississippi Delta have not been determined. We examined trends in CVD risk factors from 2001 to 2010 in the region. Longitudinal trends in prevalence of high blood pressure, high cholesterol, diabetes, obesity, physical inactivity, and current smoking were investigated using self-reported data from the Behavioral Risk Factor Surveillance System. Joinpoint regression models were used to examine annual percentage change (APC) in the prevalence of these risk factors. Overall, from 2001 to 2010, we observed significant increases in the prevalence of high cholesterol (APC, 4.22%), obesity (APC, 3.65%), and diabetes (APC, 3.54%). Among blacks, we found significant increases in the prevalence of high cholesterol (APC, 3.41%), obesity (APC, 3.48%), and diabetes (APC, 4.96%). Among whites, we found significant increases in high blood pressure (APC, 2.18%), high cholesterol (APC, 4.78%), obesity (APC, 4.18%), and physical inactivity (APC, 3.06%). We also observed a significant decrease in smoking among whites (APC, -1.99%). From 2001 to 2010, we found a significant increase in the prevalence of high cholesterol, diabetes, and obesity in the Mississippi Delta. We also observed racial differences in those prevalences.