EMAIL THIS PAGE TO A FRIEND
Neuroscience research
The NR2B antagonist, ifenprodil, corrects the l-DOPA-induced deficit of bilateral movement and reduces c-Fos expression in the subthalamic nucleus of hemiparkinsonian rats.
PMID 25697393
Abstract
The use of NR2B antagonists in Parkinsonism is still controversial. To examine their anti-parkinsonian effects, the NR2B antagonist, ifenprodil, and L-DOPA were administered together and separately in hemiparkinsonian rats (hemi-PD) that were subjected to a cylinder test. Recovery from hypoactivity was achieved by single administration of 3-7 mg/kg of L-DOPA; however, improvement in the deficit of bilateral forelimb use was not observed. When administered alone, ifenprodil had no anti-parkinsonian effects; however, combined administration of ifenprodil and 7 mg/kg of L-DOPA significantly reversed the deficit of bilateral forelimb use without adversely affecting the L-DOPA-induced improvement in motor activity. Next, in order to identify the brain area influenced by L-DOPA and ifenprodil, quantitative analysis of L-DOPA-induced c-Fos immunoreactivity was performed in various brain areas of hemi-PD following administration of L-dopa with and without ifenprodil. Among brain areas with robust c-Fos expression within the motor loop circuit in dopamine-depleted hemispheres, co-administered ifenprodil markedly attenuated L-DOPA-induced c-Fos expression in only the subthalamic nucleus (STN), suggesting that the STN is the primary target for the anti-parkinsonian action of NR2B antagonists.