EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
DBZ regulates cortical cell positioning and neurite development by sustaining the anterograde transport of Lis1 and DISC1 through control of Ndel1 dual-phosphorylation.
PMID 25698733
Abstract
Cell positioning and neuronal network formation are crucial for proper brain function. Disrupted-in-Schizophrenia 1 (DISC1) is anterogradely transported to the neurite tips, together with Lis1, and functions in neurite extension via suppression of GSK3β activity. Then, transported Lis1 is retrogradely transported and functions in cell migration. Here, we show that DISC1-binding zinc finger protein (DBZ), together with DISC1, regulates mouse cortical cell positioning and neurite development in vivo. DBZ hindered Ndel1 phosphorylation at threonine 219 and serine 251. DBZ depletion or expression of a double-phosphorylated mimetic form of Ndel1 impaired the transport of Lis1 and DISC1 to the neurite tips and hampered microtubule elongation. Moreover, application of DISC1 or a GSK3β inhibitor rescued the impairments caused by DBZ insufficiency or double-phosphorylated Ndel1 expression. We concluded that DBZ controls cell positioning and neurite development by interfering with Ndel1 from disproportionate phosphorylation, which is critical for appropriate anterograde transport of the DISC1-complex.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1784
Aminopterin, powder
C19H20N8O5
A3411
Aminopterin, powder, BioReagent, suitable for cell culture
C19H20N8O5
A5159
Aminopterin, Hybri-Max, 50 ×, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
C19H20N8O5
Y0000441
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard
C5H4N4O
77662
Hypoxanthine, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
C5H4N4O
H9377
Hypoxanthine, ≥99.0%
C5H4N4O
H9636
Hypoxanthine, powder, BioReagent, suitable for cell culture
C5H4N4O
44208
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
C6H14N2O2 · HCl
PHR1101
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C6H14N2O2 · HCl
67448
L-Lysine monohydrochloride, certified reference material, TraceCERT®
C6H14N2O2 · HCl
L0900000
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C6H14N2O2 · HCl
63031
Magnesium ion solution for ISE, 0.01 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
63042
Magnesium ion solution for ISE, 0.1 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
95660
o-Xylene, analytical standard
C8H10
294780
o-Xylene, anhydrous, 97%
C8H10
295884
o-Xylene, for HPLC, 98%
C8H10
PHR1312
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10
71383
Sodium chloride, tested according to Ph.Eur.
ClNa
71387
Sodium chloride, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
ClNa
PHR1321
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClNa
71177
Sodium standard for AAS, ready-to-use, traceable to BAM, in H2O
ClNa
T1895
Thymidine, powder, BioReagent, suitable for cell culture
C10H14N2O5
T9250
Thymidine, ≥99%
C10H14N2O5
89270
Thymidine, ≥99.0% (HPLC)
C10H14N2O5