EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Ifenprodil and SL 82.0715 are antagonists at the polyamine site of the N-methyl-D-aspartate (NMDA) receptor.
PMID 2569980
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2892
Ifenprodil (+)-tartrate salt
C42H54N2O4 · C4H6O6