EMAIL THIS PAGE TO A FRIEND
Molecular cancer therapeutics
Trifluridine Induces p53-Dependent Sustained G2 Phase Arrest with Its Massive Misincorporation into DNA and Few DNA Strand Breaks.
PMID 25700705
Abstract
Trifluridine (FTD) is a key component of the novel oral antitumor drug TAS-102, which consists of FTD and a thymidine phosphorylase inhibitor. Like 5-fluoro-2'-deoxyuridine (FdUrd), a deoxynucleoside form of 5-fluorouracil metabolite, FTD is sequentially phosphorylated and not only inhibits thymidylate synthase activity, but is also incorporated into DNA. Although TAS-102 was effective for the treatment of refractory metastatic colorectal cancer in clinical trials, the mechanism of FTD-induced cytotoxicity is not completely understood. Here, we show that FTD as well as FdUrd induce transient phosphorylation of Chk1 at Ser345, and that this is followed by accumulation of p53 and p21 proteins in p53-proficient human cancer cell lines. In particular, FTD induced p53-dependent sustained arrest at G2 phase, which was associated with a proteasome-dependent decrease in the Cyclin B1 protein level and the suppression of CCNB1 and CDK1 gene expression. In addition, a p53-dependent increase in p21 protein was associated with an FTD-induced decrease in Cyclin B1 protein. Although numerous ssDNA and dsDNA breaks were induced by FdUrd, few DNA strand breaks were detected in FTD-treated HCT-116 cells despite massive FTD misincorporation into genomic DNA, suggesting that the antiproliferative effect of FTD is not due to the induction of DNA strand breaks. These distinctive effects of FTD provide insights into the cellular mechanism underlying its antitumor effect and may explain the clinical efficacy of TAS-102.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2500182
Anti-CACNA1C antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA039796
Anti-CACNA1C antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV30161
Anti-CCNB1 antibody produced in rabbit, affinity isolated antibody
SAB2100360
Anti-CCNB1 antibody produced in rabbit, affinity isolated antibody
SAB2100378
Anti-CDC2 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100379
Anti-CDC2 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2103891
Anti-CDC2 antibody produced in rabbit, affinity isolated antibody
SAB4500051
Anti-CDC2 antibody produced in rabbit, affinity isolated antibody
SAB4503707
Anti-CDC2 antibody produced in rabbit, affinity isolated antibody
HPA003387
Anti-CDK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300390
Anti-CHEK1 (Ab-317) antibody produced in rabbit, affinity isolated antibody
AV32589
Anti-CHEK1 antibody produced in rabbit, affinity isolated antibody
SAB2104134
Anti-CHEK1 antibody produced in rabbit, affinity isolated antibody
GW21552 Anti-CHEK2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1407116
Anti-CHEK2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001878
Anti-CHEK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV30290
Anti-CHEK2 antibody produced in rabbit, affinity isolated antibody
SAB2100415
Anti-CHEK2 antibody produced in rabbit, affinity isolated antibody
SAB4500207
Anti-CHK1 antibody produced in rabbit, affinity isolated antibody
SAB4500208
Anti-CHK1 antibody produced in rabbit, affinity isolated antibody
SAB4500209
Anti-CHK1 antibody produced in rabbit, affinity isolated antibody
SAB4500210
Anti-CHK1 antibody produced in rabbit, affinity isolated antibody
SAB4500211
Anti-CHK1 antibody produced in rabbit, affinity isolated antibody
SAB3500192
Anti-Chk2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
C9108 Anti-Chk2 antibody, Mouse monoclonal, clone DCS-273, purified from hybridoma cell culture
C9233
Anti-Chk2 antibody, Mouse monoclonal, clone DCS-270, purified from hybridoma cell culture
C8831
Anti-Cyclin B1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4503501
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody
SAB4503502
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody
SAB1303610
ANTI-CYP5A1 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution